APO455838422 Apollo Scientific Ltd

3-Fluoro-2-methoxy-5-(methylthio)benzeneboronic acid

  • MDL number MFCD09878322
  • Cas Number : 957065-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42562

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=42562
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart