APO455838426 Apollo Scientific Ltd

1,2-Dichloro-3,3,3-trifluoro-2-(trifluoromethyl)propane

  • MDL number MFCD01862044
  • Cas Number : 138690-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Dichlorohexafluoroisobutane
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 78-79
Product Link https://store.apolloscientific.co.uk/product/12-dichloro-333-trifluoro-2-trifluoromethylpropane-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/12-dichloro-333-trifluoro-2-trifluoromethylpropane-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart