APO455838430 Apollo Scientific Ltd

2-(Aminomethyl)-3-(trifluoromethyl)pyridine hydrochloride

  • MDL number MFCD04117764
  • Cas Number : 1187932-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47176

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=47176
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart