APO455838443 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethyl)phenacyl bromide

  • MDL number MFCD03094506
  • Cas Number : 202664-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 15-17
Synonym 2-Bromo-2'-fluoro-5'-(trifluoromethyl)acetophenone, 2-Bromo-1-[2-fluoro-5-(trifluoromethyl)phenyl]ethan-1-one, 3-(Bromoacetyl)-4-fluorobenzotrifluoride
Purity 99
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6092

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6092
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart