APO455838448 Apollo Scientific Ltd

2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-2-(4-nitrophenyl)acetonitrile

  • MDL number MFCD00172215
  • Cas Number : 213994-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2853

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2853
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart