APO455838453 Apollo Scientific Ltd

3-Amino-8-chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD00974657
  • Cas Number : 338773-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273-275
Synonym 8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2881

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2881
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart