APO455838469 Apollo Scientific Ltd

3-Chloro-4-(trifluoromethoxy)benzenesulphonyl chloride

  • MDL number MFCD04972750
  • Cas Number : 886762-48-7
  • Molecular Weight : 295.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.59
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 80-84/1mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-trifluoromethoxybenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-trifluoromethoxybenzenesulphonyl-chloride
pictograms GHS05;GHS07
hstatements H314;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart