APO455838470 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethyl)phenacyl bromide

  • MDL number MFCD03094511
  • Cas Number : 544429-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-31
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6097

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=6097
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart