APO455838474 Apollo Scientific Ltd

2-Fluoro-5-(tributylstannyl)pyridine

  • MDL number MFCD07787406
  • Cas Number : 1025687-41-5
  • Molecular Weight : 386.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-5-(tributylstannyl)pyridine
Inchi Code 1S/C5H3FN.3C4H9.Sn/c6-5-3-1-2-4-7-5;3*1-3-4-2;/h1,3-4H;3*1,3-4H2,2H3;
Inchi Key VLMHLVKGRNNZQF-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-tributylstannylpyridine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-tributylstannylpyridine
pictograms GHS06;GHS08;GHS09
hstatements H301;H312;H319;H400
Availability details for
Quantity
  • Availability
  • Add to Cart