APO455838484 Apollo Scientific Ltd

2,2,3,3,4,4,5,5-Octafluorohexanediamide

  • MDL number MFCD00446345
  • Cas Number : 355-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-238
Synonym Octafluoroadipamide
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6454

Safety Information

signalword Danger
pcodes P301+P330+P331;P303+P361+P353
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6454
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart