APO455838485 Apollo Scientific Ltd

2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine

  • MDL number MFCD09817479
  • Cas Number : 613239-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-78
Synonym 4-[5-(Trifluoromethyl)pyridin-2-yl]benzyl chloride, 6-[4-(Chloromethyl)phenyl]-alpha,alpha,alpha-trifluoro-3-picoline
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38105

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38105
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart