APO455838486 Apollo Scientific Ltd

3,5-Difluorobenzhydrazide

  • MDL number MFCD01310810
  • Cas Number : 244022-63-7
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-170
Synonym 3,5-Difluorobenzoic acid hydrazide, 3,5-Difluorobenzohydrazide
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluorobenzhydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluorobenzhydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart