APO455838491 Apollo Scientific Ltd

4-Fluoro-N-methylbenzenesulphonamide

  • MDL number MFCD01211989
  • Cas Number : 433-14-7
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-n-methylbenzenesulphonamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-n-methylbenzenesulphonamide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart