APO455838494 Apollo Scientific Ltd

Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

  • MDL number MFCD03012932
  • Cas Number : 146137-92-0
  • Molecular Weight : 260.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methoxycarbonyl)-5-(trifluoromethyl)-1-benzothiophene, Methyl 5-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-trifluoromethylbenzobthiophene-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/methyl-5-trifluoromethylbenzobthiophene-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart