APO455838497 Apollo Scientific Ltd

4-Bromo-3-fluorobenzoyl chloride

  • MDL number MFCD09746051
  • Cas Number : 695188-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.743
Flash Point 103
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 246
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38169

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38169
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart