APO455838500 Apollo Scientific Ltd

6-(2-Fluorophenyl)-1,3,5-triazine-2,4-diamine

  • MDL number MFCD04973088
  • Cas Number : 30530-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243-247
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8775

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=8775
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart