APO455838503 Apollo Scientific Ltd

Methyl 2,5-difluoro-4-nitrobenzoate

  • MDL number MFCD08689670
  • Cas Number : 924868-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-52
Synonym 2,5-Difluoro-4-(methoxycarbonyl)nitrobenzene
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38981

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=38981
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart