APO455838508 Apollo Scientific Ltd

N-(Piperid-4-yl)-3-(trifluoromethylsulphonyl)aniline

  • MDL number MFCD01319328
  • Cas Number : 387350-88-1
  • Molecular Weight : 308.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-126
Synonym 4-3-(Trifluoromethylsulphonyl)anilinopiperidine
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-piperid-4-yl-3-trifluoromethylsulphonylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-piperid-4-yl-3-trifluoromethylsulphonylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart