APO455838509 Apollo Scientific Ltd

3-Amino-5-nitrobenzotrifluoride

  • MDL number MFCD00061229
  • Cas Number : 401-94-5
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-81
Synonym 3-Nitro-5-(trifluoromethyl)aniline, 5-Nitro-alpha,alpha,alpha-trifluoro-m-toluidine
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-5-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-5-nitrobenzotrifluoride
pictograms GHS06;GHS07
hstatements H311;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart