APO455838512 Apollo Scientific Ltd

2-Bromo-1-fluoronaphthalene

  • MDL number MFCD09864690
  • Cas Number : 317-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -4
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 164/2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39052

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39052
pictograms GHS07
hstatements H315;H319;H335;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart