APO455838513 Apollo Scientific Ltd

5-Fluoroisatoic anhydride

  • MDL number MFCD03300719
  • Cas Number : 321-69-7
  • Molecular Weight : 181.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
Inchi Code 1S/C8H4FNO3/c9-4-1-2-6-5(3-4)7(11)13-8(12)10-6/h1-3H,(H,10,12)
Inchi Key UBKGOWGNYKVYEF-UHFFFAOYSA-N

Description

Synonym 6-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione, 6-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoroisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoroisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart