APO455838514 Apollo Scientific Ltd

4-Fluoroisatoic anhydride

  • MDL number MFCD03839826
  • Cas Number : 321-50-6
  • Molecular Weight : 181.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231-232
Synonym 7-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione, 7-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoroisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoroisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart