APO455838515 Apollo Scientific Ltd

3-Fluoroisatoic anhydride

  • MDL number MFCD06796569
  • Cas Number : 174463-53-7
  • Molecular Weight : 181.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione, 8-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoroisatoic-anhydride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoroisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart