APO455838516 Apollo Scientific Ltd

5-(Bromomethyl)-2-(trifluoromethyl)pyridine

  • MDL number MFCD10697684
  • Cas Number : 108274-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-38
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47982

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47982
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart