APO455838517 Apollo Scientific Ltd

2-Chloro-3,5-difluoroaniline

  • MDL number MFCD00134427
  • Cas Number : 36556-60-2
  • Molecular Weight : 163.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26-28
Purity 97
Density 1.434
Flash Point 92
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-35-difluoroaniline

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-35-difluoroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart