APO455838518 Apollo Scientific Ltd

3-Chloro-2,4-difluorobromobenzene

  • MDL number MFCD00142590
  • Cas Number : 201849-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-chloro-2,4-difluorobenzene
Density 1.8
Flash Point >110
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2407

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2407
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart