APO455838519 Apollo Scientific Ltd

4-Chloro-3,5-difluorobromobenzene

  • MDL number MFCD00143420
  • Cas Number : 176673-72-6
  • Molecular Weight : 227.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-chloro-1,3-difluorobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-35-difluorobromobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-35-difluorobromobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart