APO455838522 Apollo Scientific Ltd

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyl diacrylate

  • MDL number MFCD00078412
  • Cas Number : 2264-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.2537
Flash Point >110
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4209

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4209
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart