APO455838523 Apollo Scientific Ltd

4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene

  • MDL number MFCD01861132
  • Cas Number : 105529-58-6
  • Molecular Weight : 259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-alpha,alpha,alpha,2-tetrafluoroanisole, 4-Bromo-2-fluorophenyl trifluoromethyl ether
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-1-trifluoromethoxybenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-1-trifluoromethoxybenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart