APO455838526 Apollo Scientific Ltd

2,4-Difluoro-5-nitrophenol

  • MDL number MFCD08443942
  • Cas Number : 113512-57-5
  • Molecular Weight : 175.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 112-120
Iupac Name 2,4-difluoro-5-nitrophenol
Inchi Code 1S/C6H3F2NO3/c7-3-1-4(8)6(10)2-5(3)9(11)12/h1-2,10H
Inchi Key SMRYCTJAGPDVEH-UHFFFAOYSA-N

Description

Synonym 2,4-Difluoro-5-hydroxynitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 112-120
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-5-nitrophenol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoro-5-nitrophenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart