APO455838527 Apollo Scientific Ltd

But-1-yl 2,3-difluorophenyl ether

  • MDL number MFCD07368738
  • Cas Number : 136239-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8951

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=8951
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart