APO455838528 Apollo Scientific Ltd

Ethyl 2-cyano-3-[4-(trifluoromethyl)phenyl]acrylate

  • MDL number MFCD00176452
  • Cas Number : 149550-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39100

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=39100
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart