APO455838532 Apollo Scientific Ltd

2,3-Dihydro-5-fluorobenzo[b]furan

  • MDL number MFCD08236758
  • Cas Number : 245762-35-0
  • Molecular Weight : 138.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 68
Iupac Name 5-fluoro-2,3-dihydro-1-benzofuran
density 1.214
Inchi Code 1S/C8H7FO/c9-7-1-2-8-6(5-7)3-4-10-8/h1-2,5H,3-4H2
Inchi Key PAVTXINZFWRQOD-UHFFFAOYSA-N

Description

Melting Point 68
Synonym 2,3-Dihydro-5-fluoro-1-benzofuran
Density 1.214
Flash Point 191
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-dihydro-5-fluorobenzobfuran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dihydro-5-fluorobenzobfuran
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart