APO455838533 Apollo Scientific Ltd

4-Fluoro-2-(hydroxymethyl)benzonitrile

  • MDL number MFCD09864693
  • Cas Number : 1000339-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point >110
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39683

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=39683
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart