APO455838537 Apollo Scientific Ltd

1-Bromo-1,1,2,2-tetrafluorobutane

  • MDL number MFCD00077466
  • Cas Number : 127117-30-0
  • Molecular Weight : 208.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point 73
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 143-144
Product Link https://store.apolloscientific.co.uk/product/1-bromo-1122-tetrafluorobutane

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-1122-tetrafluorobutane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart