APO455838538 Apollo Scientific Ltd

4-Nitrophenylsulphur pentafluoride

  • MDL number MFCD00221619
  • Cas Number : 2613-27-6
  • Molecular Weight : 249.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Synonym 4-(Pentafluorothio)nitrobenzene, 1-Nitro-4-(pentafluorosulphanyl)benzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 76-77/12mm
Product Link https://store.apolloscientific.co.uk/product/4-nitrophenylsulphur-pentafluoride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-nitrophenylsulphur-pentafluoride
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart