APO455838540 Apollo Scientific Ltd

3,4-Difluoro-2-methoxyaniline

  • MDL number MFCD07368889
  • Cas Number : 114076-35-6
  • Molecular Weight : 159.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-5,6-difluoroanisole
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-difluoro-2-methoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-difluoro-2-methoxyaniline
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart