APO455838544 Apollo Scientific Ltd

4-Hydroxy-5-phenyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole

  • MDL number MFCD08444001
  • Cas Number : 924868-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-204
Synonym 5-Phenyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-ol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39408

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39408
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart