APO455838545 Apollo Scientific Ltd

4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid

  • MDL number MFCD00013574
  • Cas Number : 1763-28-6
  • Molecular Weight : 208.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26-28
Synonym 4,4,4-Trifluoro-3-(trifluoromethyl)but-2-enoic acid
Purity 9597
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 109-110/120mm
Product Link https://store.apolloscientific.co.uk/product/444-trifluoro-3-trifluoromethylcrotonic-acid

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/444-trifluoro-3-trifluoromethylcrotonic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart