APO455838548 Apollo Scientific Ltd

3,4,5-Trifluorobenzylamine

  • MDL number MFCD00236318
  • Cas Number : 235088-69-4
  • Molecular Weight : 161.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3,4,5-Trifluorophenyl)methylamine
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/345-trifluorobenzylamine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/345-trifluorobenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart