APO455838551 Apollo Scientific Ltd

5'-Chloro-2'-fluoroacetophenone

  • MDL number MFCD03094519
  • Cas Number : 541508-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-42
Synonym 1-(5-Chloro-2-fluorophenyl)ethan-1-one
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2616

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2616
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart