APO455838557 Apollo Scientific Ltd

6-(Trifluoromethyl)nicotinamidine hydrochloride

  • MDL number MFCD01862652
  • Cas Number : 221313-11-7
  • Molecular Weight : 225.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-193
Synonym 6-(Trifluoromethyl)pyridine-3-carboxamidine hydrochloride, 6-(Trifluoromethyl)pyridine-3-carboximidamide hydrochloride, 5-Carbamimidoyl-2-(trifluoromethyl)pyridine hydrochloride
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-trifluoromethylnicotinamidine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-trifluoromethylnicotinamidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart