APO455838558 Apollo Scientific Ltd

Trifluoropyruvaldehyde, 20% aqueous solution

  • MDL number MFCD03092983
  • Cas Number : 91944-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Oxo-3,3,3-trifluoropropanal, Trifluoromethylglyoxal
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12303

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12303
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart