APO455838559 Apollo Scientific Ltd

2-[4-(Trifluoromethyl)phenyl]pyrrolidine

  • MDL number MFCD01631837
  • Cas Number : 298690-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-4-trifluoromethylphenylpyrrolidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-trifluoromethylphenylpyrrolidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart