APO455838560 Apollo Scientific Ltd

3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile

  • MDL number MFCD00664066
  • Cas Number : 80194-70-3
  • Molecular Weight : 206.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-2-cyano-5-(trifluoromethyl)pyridine, 3-Chloro-5-(trifluoromethyl)picolinonitrile
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 90/11mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-trifluoromethylpyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-trifluoromethylpyridine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart