APO455838563 Apollo Scientific Ltd

1-[4-(Trifluoromethyl)phenyl]propan-1-ol

  • MDL number MFCD07773046
  • Cas Number : 67081-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.2
Flash Point 104
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11264

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=11264
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart