APO455838565 Apollo Scientific Ltd

2-Chloro-6-(trifluoromethyl)benzaldehyde

  • MDL number MFCD06660231
  • Cas Number : 60611-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11251

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=11251
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart