APO455838568 Apollo Scientific Ltd

1-Bromo-5-fluoronaphthalene

  • MDL number MFCD09258916
  • Cas Number : 315-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Purity 97
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 85-90/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33407

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33407
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart