APO455838570 Apollo Scientific Ltd

3,5-Dibromo-2-fluoropyridine

  • MDL number MFCD07368619
  • Cas Number : 473596-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/35-dibromo-2-fluoropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dibromo-2-fluoropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart