APO455838574 Apollo Scientific Ltd

2-Fluoro-3-methylaniline

  • MDL number MFCD06410915
  • Cas Number : 1978-33-2
  • Molecular Weight : 125.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Amino-2-fluorotoluene, 2-Fluoro-m-toluidine
Density 1.1
Flash Point 80
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-methylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-methylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart